missing translation for 'onlineSavingsMsg'
Learn More

Luminol, TRC

Product Code. 30481435
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481435 50 g 50g
30475599 1 g 1g
30469193 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481435 Supplier Toronto Research Chemicals Supplier No. TRCL47445050G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8 H7 N3 O2
Molecular Weight (g/mol) 177.16
Synonym 1,4-Phthalazinedione, 5-amino-2,3-dihydro-, 5-Amino-2,3-dihydro-1,4-phthalazinedione, 3-Aminophthalhydrazide, 3-Aminophthalic acid hydrazide, 3-Aminophthalic hydrazide, 5-Amino-1,4-dihydroxyphthalazine, Diogenes reagent, Luminol, NSC 5064
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES Nc1cccc2C(=O)NNC(=O)c12

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H7N3O2/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12/h1-3H,9H2,(H,10,12)(H,11,13)
Formula Weight 177.0538
Chemical Name or Material Luminol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.