missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Luminol and oxidizing solutions (each bottle)

Product Code. 15474059
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474059 250 mL 250mL
15484059 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474059 Supplier Thermo Scientific Chemicals Supplier No. J61462.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For chemiluminecent detection of HRP-based Western blotting systems

Used by biologists in cellular assays for the detection of copper, iron, and cyanides, as well as of specific proteins by western blot. For chemiluminecent HRP-based detection of western blotting systems. Microestimation of glucose and glucose oxidase using enzyme-induced chemiluminescence6,7 - Inhibitor of poly ADP Ribose Synthase8 - Demonstrations of the phenomenon of chemiluminescence, using different sensitizers to produce different colors of light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Luminol and oxidizing solutions (each bottle)
Solubility Information It is soluble in most polar organic solvents but is insoluble in water.
Quantity 250 mL
Sensitivity Light sensitive
Physical Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.