missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminex™ 200™ Instrument System

Product Code. 30245461
Change view
Click to view available options
Quantity:
1 system
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30245461 1 system Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245461 Supplier Invitrogen™ Supplier No. APX10031

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

The Luminex 200 Instrument System sets the standard for multiplexing, providing the ability to perform up to 100 different tests in a single reaction volume on a flow cytometry-based platform.

The Luminex 200 Instrument System sets the standard for multiplexing, providing the ability to perform up to 100 different tests in a single reaction volume on a flow cytometry-based platform. The Luminex 200 system is compatible with ProcartaPlex multiplex immunoassays and QuantiGene Plex multiplex gene expression assays.

Features of the Luminex system include:

  • Multiplexing: up to 80 analytes per sample using magnetic microspheres
  • Dynamic range (typical): ≥3.5 logs
  • Read time: 96-well plate in ≤45 min (up to 12,800 tests/hour)
  • Daily start-up/shut-down: ≤30 min

Available options

  • Software: extra seat licenses, Basic Security Module, 21 CFR part 11 Module, Automation Module, LIS/LIMS Module
  • Service: service contracts, IQ/OQ

Luminex xMAP technology

The open architecture of Luminex xMAP technology uses imaging, microspheres, digital signal processing, and traditional chemistry—combining proven technologies in a unique way.

Advantages of Luminex xMAP technology include:

  • Helps reduce costs and labor through multiplexing
  • Smaller sample size requirements compared to single result assays
  • Enables fast, reproducible results from favorable kinetics of liquid bead array approach
  • Broad coverage of applications, including protein expression and gene expression profiling

How Luminex xMAP technology works

Sets of magnetic microspheres are color-coded with combinations of red and infrared fluorophores. Each bead set is coated with a reagent specific to a particular bioassay, allowing the capture and detection of several specific analytes from a single sample. Optics in the compact analyzer excite the internal dyes that identify each microsphere as well as any reporter dye captured, providing quantitation of individual analytes on each microsphere. The analyzer reads many beads from each set, enabling rapid and precise results for several targets within a single sample.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Luminex 200 instrument
• xMAP Sheath Fluid Sheath Delivery System
• X-Y Platform for 96-Well Plates
• xMAP Sheath Fluid
• Sample Probe Needle
• 1 L Sheath Bottle
• 1.2 L Waste Bottle
• Sample Probe Height Adjustment Kit
• Automated Maintenance Plate
• Heater Block (96-well plate)
• Documentation
• Desktop PC with Windows 10; Dell monitor, keyboard, and mouse; xPONENT Software (3 seats), and 90-day trial of Security, 21 CFR Part 11, Automation and LIS modules

Shipped separately:
• Luminex 200 Calibration Kit
• Luminex 200 Performance Verification Kit

Shipping Condition Room Temperature
Throughput Up to 100 Tests per Hour
Quantity 1 system

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.