missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Luminescence Test Plate LB 9515

Product Code. 31777432 Shop All Berthold Technologies Products
Change view
Click to view available options
Type:
QC Microplate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
31777432 QC Microplate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777432 Supplier Berthold Technologies Supplier No. 4010512
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easy-to-use luminescence test plate for checking microplate luminometer performance and simulating sample light emission.

Highlights

  • Verifies and adjusts measurement position for microplate luminometers (96-well and 384-well formats)
  • Enables verification of crosstalk, detector function, and detector stability
  • Uses diffusers to simulate light emission characteristics of samples
  • Supplied in a transport case for use across multiple instrument locations

Technical Details

Available Positions

  • 96-well position
  • 384-well position
  • Overload position

Parameters Tested

  • Measurement position (96-well format)
  • Crosstalk (96-well format)
  • Detector stability
  • Measurement position (384-well format)
  • Crosstalk (384-well format)
  • Detector function (overload position)

Compatibility

  • Centro series
  • Mithras series
  • Orion II
  • Tristar series

Specifications

Type QC Microplate
Light Source LED
Detection Method Luminescence
Color Black
For Use With (Application) Quality Control
Voltage 6 V
For Use With (Equipment) Lumat LB 9510 Tube Luminometer
Includes Charger, Transport Case, Test Plate
Certifications/Compliance CE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.