missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luminescence Enhancer For Solution Assays, Emerald™ Enhancer
Description
The Emerald™ chemiluminescence enhancer
is a concentrated reagent suitable for use with chemiluminescent substrates (AMPPD™, CSPD™, Galacton™, Glucon™, or Glucuron™).
For Research Use Only. Not for use in diagnostics procedures.
is a concentrated reagent suitable for use with chemiluminescent substrates (AMPPD™, CSPD™, Galacton™, Glucon™, or Glucuron™).
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Product Type | Luminescence Enhancer |
| Content And Storage | Store the product at 4°C. |
| Format | Bottle(s) |
| Form | Liquid |
| Shipping Condition | Room Temperature |
| Substrate Type | AP (Alkaline Phosphatase) Substrate |
| Substrate | Alkaline Phosphatase |
| Product Line | Emerald, NovaBright |
| Quantity | 2500 assays |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?