missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Luminescence Enhancer For Membrane Blotting Assays, Nitro-Block™ Enhancer
Shop All Applied Biosystems Products
Click to view available options
Quantity:
20 mL
Unit Size:
Each
Description
This concentrated Nitro-Block™ Enhancer can be used with CSPD™ chemiluminescent substrate to increase the alkaline phosphatase signal for protein or nucleic acid blots.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Description | Nitro-Block™ Luminescence Enhancer, 20mL, 20X Concentration, Liquid, Alkaline Phosphatase Substrate, For Membrane Blotting Assays, Used For Protein Assays and Analysis, 4°C Storage Temperature, Room Temperature |
| Concentration | 20 x |
| Formulation | Liquid |
| For Use With (Application) | Protein assays and analysis |
| Sample Volume | 20mL |
| Content And Storage | 4°C |
| Product Type | Nitro-Block™ Luminescence Enhancer |
| Quantity | 20 mL |
| Format | Bottles |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction