missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Luminata Forte ELISA HRP Substrate
Used in Western blotting; dot/slot blotting
Brand: Merck ELLUF0100
295.03 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Detection Methods: Chemiluminescence
- Key Applications: ELISA
- Chemiluminescent: HRP detection reagent
- For use on immunoblots
Specifications
100mL | |
2° to 8°C |