missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Luminata Crescendo ELISA HRP Substrate
Shop All Merck Products
Click to view available options
Quantity:
100 mL
200 mL
Unit Size:
100mL
200mL
Description
- Detection Methods: Chemiluminescence
- Key Applications: ELISA
- Chemiluminescent: HRP detection reagent
- For use on immunoblots
Specifications
Specifications
| Quantity | 100 mL |
| Content And Storage | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction