missing translation for 'onlineSavingsMsg'
Learn More

Merck Luminata Crescendo ELISA HRP Substrate

Product Code. 12561209 Shop All Merck Products
Change view
Click to view available options
Quantity:
100 mL
200 mL
Unit Size:
100mL
200mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12561209 100 mL 100mL
11357630 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12561209 Supplier Merck Supplier No. ELLUR0100
  • ONLINE EXCLUSIVE PRICE
    £312.00
    £278.53 / 100mL
    Save £33.47
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used on immunoblots as chemiluminescent substrate

  • Detection Methods: Chemiluminescence
  • Key Applications: ELISA
  • Chemiluminescent: HRP detection reagent
  • For use on immunoblots
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.