missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminaris Color HiGreen qPCR Master Mix, high ROX

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

Brand:  Thermo Scientific™ K0362

Code : 88

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 13213179

  • £487.00 / 500 reactions
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • Luminaris Color HiGreen High ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer with a blue dye. The master mix contains high concentration of ROX passive reference

Description

Description

Thermo Scientific™ Luminaris Color HiGreen Master Mixes are ready-to-use qPCR master mixes with color and premixed UDG optimized for SYBR Green chemistry using standard cycling protocols.

  • Blue master mix and Yellow Sample Buffer for easy pipetting
  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
Specifications

Specifications

Contains:
• 4 x 1.25 mL Luminaris Color HiGreen High ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer with a blue dye; sufficient for 500 x 20 μL reactions. The master mix contains high concentration of ROX passive reference dye for specific real-time platforms.
• 1 x 1.25 mL 40X Yellow Sample Buffer
• 4 x 1.25 mL Nuclease-Free Water

Store at -20°C.
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
qPCR Master Mix
Gene Expression
2 X
DNA, dsDNA
SYBR
qPCR
Standard
Luminaris
500 Reactions
2X
500 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.