missing translation for 'onlineSavingsMsg'
Learn More

Lumiflavin, TRC

Product Code. 17949844
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949844 100 mg 100mg
17918224 5 mg 5mg
17907634 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17949844 Supplier Toronto Research Chemicals Supplier No. TRCL473900100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1088-56-8
Molecular Formula C13H12N4O2
Molecular Weight (g/mol) 256.26
Synonym Benzo[g]pteridine-2,4(3H,10H)-dione, 7,8,10-trimethyl-, Benzo[g]pteridin-2(10H)-one, 4-hydroxy-7,8,10-trimethyl- (7CI), Lumiflavine (6CI,8CI), 7,8,10-Trimethylbenzo[g]pteridine-2,4(3H,10H)-dione, 7,8,10-Trimethylisoalloxazine, Lumiflavin, Lumilactoflavin
IUPAC Name 7,8,10-Trimethylbenzo[g]pteridine-2,4(3H,10H)-dione
SMILES CN1C2=NC(=O)NC(=O)C2=Nc3cc(C)c(C)cc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H12N4O2/c1-6-4-8-9(5-7(6)2)17(3)11-10(14-8)12(18)16-13(19)15-11/h4-5H,1-3H3,(H,16,18,19)
Formula Weight 256.096
Percent Purity >95%
Chemical Name or Material Lumiflavin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.