missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lumican Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10677-1-AP-150UL
This item is not returnable.
View return policy
Description
LUM is a member of the small leucine-rich proteoglycan (SLRP) family that includes decorin, biglycan, fibromodulin, keratocan, epiphycan, and osteoglycin. In these bifunctional molecules, this protein moiety binds collagen fibrils and the highly charged hydrophilic glycosaminoglycans regulate interfibrillar spacings. The protein is the major keratan sulfate proteoglycan of the cornea but is also distributed in interstitial collagenous matrices throughout the body. Lumican may regulate collagen fibril organization and circumferential growth, corneal transparency, and epithelial cell migration and tissue repair.Specifications
| Lumican | |
| Polyclonal | |
| Unconjugated | |
| LUM | |
| KSPG lumican, LDC, LUM, lumican, SLRR2D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17022, 4060, 81682 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P51884, P51885, P51886 | |
| Lum | |
| Lumican Fusion Protein Ag1106 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction