missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lumateperone tosylate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Lumateperone (ITI-007) tosylate is a 5-HT2A receptor antagonist (Ki = 0.54 nM), a partial agonist of presynaptic D2 receptors and an antagonist of postsynaptic D2 receptors (Ki = 32 nM), and a dopamine D1 receptor modulator. Lumateperone tosylate has anticancer activity and can also be used in studies of psychiatric disorders such as schizophrenia.
Specifications
Specifications
| Chemical Name or Material | Lumateperone tosylate |
| CAS | 1187020-80-9 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C31H36FN3O4S |
| Quantity | 5 mg |
| Synonym | ITI-007 tosylate |
| Solubility Information | DMSO : 100 mg/mL (176.77 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.77 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(C1=CC=C(F)C=C1)CCCN2CC[C@@](N3CCN(C)C4=C3C5=CC=C4)([H])[C@@]5([H])C2.O=S(C6=CC=C(C)C=C6)(O)=O |
| Molecular Weight (g/mol) | 565.7 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction