Learn More
Invitrogen™ LUM Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA514571
Product Code. 10374240
  • £479.00 / 400µL
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
LUM is a member of the small leucine-rich proteoglycan (SLRP) family that includes decorin, biglycan, fibromodulin, keratocan, epiphycan, and osteoglycin. In these bifunctional molecules, this protein moiety binds collagen fibrils and the highly charged hydrophilic glycosaminoglycans regulate interfibrillar spacings. The protein is the major keratan sulfate proteoglycan of the cornea but is also distributed in interstitial collagenous matrices throughout the body. Lumican may regulate collagen fibril organization and circumferential growth, corneal transparency, and epithelial cell migration and tissue repair.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LUM
Polyclonal
Unconjugated
LUM
keratan sulfate proteoglycan lumican; KSPG lumican; Lcn; Ldc; Lum; Lumican; Lumican precursor; lumican proteoglycan; SLRR2D
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
4060
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P51884
LUM
KLH conjugated synthetic peptide between 198-227 amino acids from the central region of human LUM.
400 μL