missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Lugol’s Iodine Stain, Concentrated

Product Code. 12996307
Change view
Click to view available options
Quantity:
100mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12996307 100mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996307 Supplier Pro-Lab Diagnostics™ Supplier No. PL.8010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a mordant in Gram staining for easily identifying gram positive organisms. Pro-Lab Diagnostics™ Lugol’s Iodine Stain, Concentrated is a 10x concentrated microbiological stain manufactured to the highest standards, according to all current regulatory requirements, and CE labelled.

Pro-Lab Diagnostics™ stains are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all stains offer excellent performance characteristics.

  • 10X concentrated – 100mL is diluted to 1L of ready-to-use stain
  • Apply after staining with crystal violet, but before ethanol
  • Stains the peptidoglycan of gram positive organisms

Specifications

Solution Type Concentrated
Type Lugol's Iodine
Quantity 100mL
Certifications/Compliance CE marked (98/78/EC)
Format Liquid
For Use With (Application) Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria
Color Dark Brown
Composition Ethanol 25 – <50%, Potassium iodide 10 – <25%, Iodine 5 – <10%, Methanol 2.5 – <5%
Storage Requirements Keep at temperature not exceeding 20°C
Stain Type Gram
Volume (Metric) 100 mL (Yields 1 L Ready-to-Use Stain)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lugols Iodine Concentrate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P364-And wash it before reuse.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, potassium iodide, iodine, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.