missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Lugol’s Iodine Stain
Description
Pro-Lab Diagnostics™ stains are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all stains offer excellent performance characteristics.
- Ready-to-use stain in convenient and economical sizes
- Apply after staining with crystal violet, but before ethanol
- Stains the peptidoglycan of gram positive organisms
Specifications
Specifications
| Solution Type | Reagent |
| Type | Lugol's Iodine |
| Quantity | 1L |
| Certifications/Compliance | (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark |
| Format | Liquid |
| For Use With (Application) | Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria |
| Color | Dark brown |
| Composition | Ethanol 2.5 – <5%, Potassium iodide 1 – <2.5%, Iodine 0.5 – <1%, Methanol 0.25 – <0.5% |
| Storage Requirements | Store in a cool, well-ventilated place |
| Stain Type | Gram Stain |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?