Learn More
Value Plastics Luer Thread Fittings, Elbow
Shop All Cole-Parmer Products
Click to view available options
Inlet:
Female Luer Thread
Male Luer to Female Luer
Male Luer, Female Luer Thread
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Inlet | Female Luer Thread |
| Material (Seal) | Polypropylene |