Description
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Inlet | Male Luer to Female Luer |
| Material (Seal) | Polypropylene |