missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Tuberculin Luer Syringes

Product Code. 12732637
Change view
Click to view available options
Volume (Metric):
5 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12732637 5 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12732637 Supplier B Braun™ Supplier No. 4606051V

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Use this syringe type for tuberculin injections. the B Braun™ Tuberculin Leur Syringes are an essential tool for this type of patient medical need.

  • Minimal silicone residue
  • Positive plunger stop
  • 5mL
  • Weight: 1.0kg

Specifications

Material Polyisoprene Rubber (Tip), Polypropylene (Barrel), Polypropylene (Plunger)
Includes Plunger
Product Type Tuberculin Syringe
Quantity 100/Pk.
Packaging 100 Pack
Volume (Metric) 5 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.