Learn More
3M™ Particulate Respirator, 8300 Series
Product Code. 12902475
Click to view available options
Filter Type:
FFP1
FFP2
FFP3
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 10
Pack of 80
Product Code. 12902475
Brand: 3M™ 7100010817
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide comfortable, effective respiratory protection against dust particles and/or non-volatile liquid particles. The 3M 8300 Series Particulate Respirator gives effective filtration with low breathing resistance for consistent high-quality performance.
Provide comfortable, effective respiratory protection for use in industries where workers will be exposed to dust particles and/or non-volatile liquid particles.
  • Tested and certified to AS/NZS 1716:2003
  • Robust outer shell improves collapse resistance
  • M-Noseclip adjusts easily for fewer pressure points and greater comfort on the nose
  • Braided headbands improve comfort on the neck, face, and head with a secure feel
  • 3M™ Advanced Electret Filter Material gives effective filtration with low breathing resistance for consistent high-quality performance
  • The super-soft, cushioned inner lining helps workers stay comfortable
  • Models that include the Cool Flow™ exhalation valve offer improved comfort in hot humid environments and/or where work is hard and physical
  • Colored headbands for easy identification: yellow for P1, blue for P2
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AS/NZS 1716:2003; European Standard EN149:2001 + A1:2009 NR
Filter Type FFP2
Material Thermoplastic Elastomer
Product Type General Purpose Particulate Respirator
With Exhalation Valve Yes
For Use With (Application) Industries where workers will be exposed to dust particles or non-volatile liquid particles
Latex Free Yes
Model 8322
Nosepiece Type Aluminum clip
Sterility Non-sterile
FDA Cleared Surgical Mask No