Learn More
Masterflex™ Polycarbonate Luer Fittings
Description
Both straight and elbow connectors are available. Hose barbs are available from 1/16 to ¼ in. with either male or female luer connections.
Choose from several different barb styles:
Choose from several different barb styles:
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
- Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Specifications
Specifications
| Description | Female x Male Lock, Rotating |
| Material | Polycarbonate |
| Sterility | Non-sterile |
| Product Type | Luer Fitting |