Description
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards. Constructed of ADCF polypropylene
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer Thread, Male Slip Luer |
| Width (English) | 29/32 in. |