missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luciferase Antibody (Luci 21 1-107), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-307B
This item is not returnable.
View return policy
Description
Luciferase Monoclonal specifically detects Luciferase in Firefly samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Luciferase | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Firefly | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Luci 21 1-107 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
| This Luciferase Antibody (Luci 21 1-107) was developed against luciferase protein from Photinus pyralis (North American firefly). [UniProt# P08659]. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins | |
| This Luciferase Antibody (Luci 21 1-107) is specific for Luciferase, recognizing a peptide consisting of the first 258 amino acids. Further epitope mapping has not been done at this time. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction