missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luciferase Antibody, DyLight 680, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1677FR
This item is not returnable.
View return policy
Description
Luciferase Polyclonal antibody specifically detects Luciferase in Firefly samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Luciferase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Multi-step | |
| RUO | |
| Firefly | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| ec 1.13.12.7, firefly luciferase, Fluc, LuC, luciferin 4 monooxygenase, Luciferin 4-monooxygenase | |
| Luciferase [Photinus pyralis (Firefly)] | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction