missing translation for 'onlineSavingsMsg'
Learn More

Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals

Catalog Number 11477532
204.60 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.


Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
67769-47-5
C13H9Li2N5O9S2
457.241
RPKCZJYDUKVMGF-UHFFFAOYSA-L
6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
124220108
CHEBI:52106
dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
[Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]
This item is not returnable. View return policy
67769-47-5
C13H9Li2N5O9S2
457.241
RPKCZJYDUKVMGF-UHFFFAOYSA-L
6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
124220108
CHEBI:52106
dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
[Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]
Assay Percent Range >95%
Formula Weight 457.2
Percent Purity ≥99%
Color Orange to Yellow
Quantity 25 mg
Physical Form Powder
Chemical Name or Material Lucifer yellow CH, Dilithium salt, For HPLC analysis
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'