missing translation for 'onlineSavingsMsg'
Learn More

Lucidin, MedChemExpress

Product Code. 16486778
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486778 10 mg 10mg
16469327 5 mg 5mg
16452857 50 mg 50mg
16406906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486778 Supplier Medchem Express Supplier No. HY15733
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lucidin (NSC 30546) is a natural component of Rubia tinctorum L.

Lucidin (NSC 30546) is a natural component of Rubia tinctorum L.

Specifications

Chemical Name or Material Lucidin
CAS 478-08-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 10 mg
Synonym NSC 30546
Solubility Information DMSO : 6.67 mg/mL (24.68 mM; Need ultrasonic)
SMILES O=C1C2=C(C=CC=C2)C(C3=CC(O)=C(CO)C(O)=C13)=O
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 97.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.