missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LUC7L3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554125
This item is not returnable.
View return policy
Description
Immunogen sequence: KQMEVCEVCG AFLIVGDAQS RVDDHLMGKQ HMGYAKIKAT VEELKEKLRK RTEEPDRDER LKKEKQEREE REKERERERE ERER Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Binds cAMP regulatory element DNA sequence. May play a role in RNA splicing.
Specifications
| LUC7L3 | |
| Polyclonal | |
| Unconjugated | |
| LUC7L3 | |
| 3300001P08Rik; cAMP regulatory element-associated protein 1; cisplatin resistance associated overexpressed protein; cisplatin resistance-associated overexpresse protein-like protein; cisplatin resistance-associated overexpressed protein; cisplatin resistance-associated-overexpressed protein; CRA; CREAP1; CREAP-1; CRE-associated protein 1; Crop; FLJ11063; hLuc7A; LUC7 like 3 pre-mRNA splicing factor; Luc7A; LUC7L3; LUC7-like 3; LUC7-like 3 (S. cerevisiae); LUC7-like 3 pre-mRNA splicing factor; luc7-like protein 3; O48; OA48-18; okadaic acid-inducible phosphoprotein OA48-18; RGD1307981; zgc:136380; zgc:158165 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360602, 51747, 67684 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95232, Q5SUF2 | |
| LUC7L3 | |
| Recombinant protein corresponding to Human LUC7L3. Recombinant protein control fragment (Product #RP-92020). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur