missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LUC7L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562446
This item is not returnable.
View return policy
Description
Immunogen sequence: RSRHSSRDRS RERSKRRSSK ERFRDQDLAS CDRDRSSRDR SPRDRDRKDK KRSYESANGR SEDRRSSEER EAG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene encodes a protein that contains a C2H2-type zinc finger, coiled-coil region and arginine, serine-rich (RS) domain. A similar protein in mouse interacts with sodium channel modifier 1, and the encoded protein may be involved in the recognition of non-consensus splice donor sites in association with the U1 snRNP spliceosomal subunit. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| LUC7L2 | |
| Polyclonal | |
| Unconjugated | |
| LUC7L2 | |
| 4930471C18Rik; AA522013; AU015269; CGI59; CGI-59; Cgi74; CGI-74; CGI-74 homolog; CGI-74-like SR-rich; LUC7 like 2, pre-mRNA splicing factor; LUC7B2; LUC7L2; LUC7-like 2; LUC7-like 2 (S. cerevisiae); LUC7-like 2 pre-mRNA splicing factor; Putative RNA-binding protein Luc7-like 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y383 | |
| LUC7L2 | |
| Recombinant protein corresponding to Human LUC7L2. Recombinant protein control fragment (Product #RP-95650). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction