missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LUC7L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567040
This item is not returnable.
View return policy
Description
Immunogen sequence: PPDWRLESSNG KMASRRSEEK EAGEI Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
May bind to RNA via its Arg/Ser-rich domain.
Specifications
| LUC7L | |
| Polyclonal | |
| Unconjugated | |
| Luc7l | |
| 1810045C04Rik; 2410018D03Rik; hLuc7B1; Luc7; Luc7 homolog (S. cerevisiae)-like; LUC7 like; LUC7B1; Luc7l; LUC7L1; LUC7-like; LUC7-like (S. cerevisiae); Putative RNA-binding protein Luc7-like 1; putative SR protein LUC7B1; sarcoplasmic reticulum protein LUC7B1; SR+89 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55692 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQ29 | |
| Luc7l | |
| Recombinant Human LUC7L. Recombinant protein control fragment (Product #RP-105470). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction