missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LUC7L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | LUC7L |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274732
|
Novus Biologicals
NBP2-56401 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615848
|
Novus Biologicals
NBP2-56401-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LUC7L Polyclonal specifically detects LUC7L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LUC7L | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 55692 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PPDWRLESSNGKMASRRSEEKEAGEI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ10231, hLuc7B1, Luc7, LUC7 (S. cerevisiae)-like, LUC7B1, LUC7L1, LUC7-LIKE, LUC7-like (S. cerevisiae), putative RNA-binding protein Luc7-like 1, Putative SR protein LUC7B1, sarcoplasmic reticulum protein LUC7B1, SR+89 | |
| LUC7L | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title