missing translation for 'onlineSavingsMsg'
Learn More

LUC7L Antibody (2D10), Novus Biologicals™

Product Code. 18351189 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351189 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351189 Supplier Novus Biologicals Supplier No. H00055692M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LUC7L Monoclonal antibody specifically detects LUC7L in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUC7L
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060502
Gene Alias FLJ10231, hLuc7B1, Luc7, LUC7 (S. cerevisiae)-like, LUC7B1, LUC7L1, LUC7-LIKE, LUC7-like (S. cerevisiae), putative RNA-binding protein Luc7-like 1, Putative SR protein LUC7B1, sarcoplasmic reticulum protein LUC7B1, SR+89
Host Species Mouse
Immunogen LUC7L (NP_060502, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSAQAQMRALLDQLMGTARDGDETRQRVKFTDDRVCKSHLLDCCPHDILAGTRMDLGECTKIHDLALRADYEIASKERDLFFELDAMDHLESFIAECDRRTELAKKRLAE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55692
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.