missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lubricin/PRG4 Monoclonal antibody specifically detects Lubricin/PRG4 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | Lubricin/PRG4 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1D5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005798 |
| Gene Alias | bG174L6.2, bG174L6.2 (MSF: megakaryocyte stimulating factor ), CACP, FLJ32635, JCAP, lubricin, megakaryocyte stimulating factor, Megakaryocyte-stimulating factor, MSFproteoglycan 4, (megakaryocyte stimulating factor, articular superficial zoneprotein, camptodactyly, arthropathy, coxa vara, pericarditis syndrome), proteoglycan 4, Superficial zone proteoglycan, SZPHAPOarticular superficial zone protein |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?