missing translation for 'onlineSavingsMsg'
Learn More

Lubiprostone, MedChemExpress

Product Code. 16471038
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471038 5 mg 5mg
16452259 1 mg 1mg
16434447 50 mg 50mg
16418578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471038 Supplier Medchem Express Supplier No. HYB0679
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lubiprostone(SPI-0211;RU0211) is a gastrointestinal agent used for the treatment of idiopathic chronic constipation.

Lubiprostone(SPI-0211

Specifications

Chemical Name or Material Lubiprostone
Physical Form Solid
CAS 136790-76-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H32F2O5
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym RU-0211 SPI-0211
Solubility Information DMSO : 100 mg/mL (256.11 mM; Need ultrasonic)
SMILES O=C(C(F)(F)CCCC)CC[C@@H]([C@@H](C1)O)[C@H](C1=O)CCCCCCC(O)=O
Molecular Weight (g/mol) 390.46
Formula Weight 390.46
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.