missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTV1 Homolog Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556372
This item is not returnable.
View return policy
Description
Immunogen sequence: ATGEEEGMDI QKSENEDDSE WEDVDDEKGD SNDDYDSAGL LSDEDCMSVP GKTHRAIADH LFWSEETKSR FTEYSM Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 49%.
LTV1 gene ontology annotations related to this gene include ribosomal small subunit biogenesis; ribosomal small subunit export from nucleus.
Specifications
| LTV1 Homolog | |
| Polyclonal | |
| Unconjugated | |
| LTV1 | |
| 2610020N02Rik; C6orf93; dJ468K18.4; Ltv1; LTV1 homolog; LTV1 homolog (S. cerevisiae); LTV1 ribosome biogenesis factor; protein LTV1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GA3 | |
| LTV1 | |
| Recombinant protein corresponding to Human LTV1 Homolog. Recombinant protein control fragment (Product #RP-94841). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction