missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTK (TYK1) Recombinant Human Protein
LTK (TYK1) is a member of the ros/insulin receptor family of kinases.
£434.00 - £6900.00
Specifications
Accession Number | NP_002335.2 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 73.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10392383
|
Invitrogen™
PV4651 |
10μg |
£434.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438653
|
Invitrogen™
PR8108A |
100μg |
£1602.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448743
|
Invitrogen™
PV4652 |
1mg |
£6900.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LTK (TYK1) is involved in cell growth and differentiation.
Specifications
NP_002335.2 | |
73.7kDa | |
RUO - research use only | |
ALK Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
LTK (TYK1) | |
Human | |
GST Tag |