missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA130314
This item is not returnable.
View return policy
Description
Positive Control: Placenta lysate.
LTK (TYK1), ALK and Ros have been identified as receptor tyrosine kinases having sequence similarity to the Insulin receptor subfamily of kinases. LTK, leukocyte tyrosine kinase, is expressed in murine B-lymphocyte precursors and has also been found in forebrain neurons. ALK, anaplastic lymphoma kinase, is normally highly expressed specifically in the nervous system. A truncated form containing the catalytic domain of ALK is expressed as the result of a translocation occurring in many non-Hodgkin's lymphomas. The c-Ros gene was originally identified in mutant form as an oncogene. Ros is normally expressed in a small number of epithelial cell types and may play a role in epithelial development.
Specifications
| LTK | |
| Polyclonal | |
| Unconjugated | |
| LTK | |
| EC 2.7.1.112; EC 2.7.10.1; leukocyte receptor tyrosine kinase; leukocyte tyrosine kinase; leukocyte tyrosine kinase receptor; Ltk; Protein tyrosine kinase 1; TYK1; tyrosine kinase-1 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4058 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P29376 | |
| LTK | |
| KLH conjugated synthetic peptide between 42-72 amino acids from the N-terminal region of human LTK. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction