missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LTC4S Polyclonal Antibody
GREENER_CHOICE

Product Code. 17732824
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17732824 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17732824 Supplier Invitrogen™ Supplier No. PA5121425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

Catalyzes the conjugation of leukotriene A4 with reduced glutathione to form leukotriene C4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTC4S
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LTC4S
Gene Accession No. Q16873
Gene Alias Leukotriene C4 synthase; Leukotriene-C(4) synthase; LTC4 synthase; LTC4S
Gene Symbols LTC4S
Host Species Rabbit
Immunogen Recombinant Human Leukotriene C4 synthase protein (36-51aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4056
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.