missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTBR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586436
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
Specifications
| LTBR | |
| Polyclonal | |
| Unconjugated | |
| Ltbr | |
| AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17000, 297604, 4055 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P36941, P50284 | |
| Ltbr | |
| Synthetic peptide, corresponding to amino acids 20-60 of Human LTbetaR. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction