Learn More
Invitrogen™ LTBR Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547040
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) LT beta R is observed, and less than 1% cross-reactivity with recombinant mouse (rm) 4-1BB, rmCD27, rmOPG, rmRANK, rmCD30, rmCD40, rhDR6, rmTNF RI, rmTNF RII, rmEDAR, rmFas and rmGITR is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
Specifications
| LTBR | |
| Polyclonal | |
| Unconjugated | |
| Ltbr | |
| AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 17000 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P50284 | |
| Ltbr | |
| Mouse myeloma cell line NS0-derived recombinant mouse Lymphotoxin beta R/TNFRSF3 Ser28-Pro218. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.