missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTB4R Recombinant Rabbit Monoclonal Antibody (24GB50)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557050
This item is not returnable.
View return policy
Description
LTB4R Recombinant Monoclonal Antibody for Western Blot, ICC/IF
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.
Specifications
| LTB4R | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88855, Q9R0Q2 | |
| LTB4R, Ltb4r1 | |
| A synthesized peptide derived from human LTB4R (aa 322-352). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 24GB50 | |
| Unconjugated | |
| Ltb4r1 | |
| BLT; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4 receptor; Ltb4r; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16995, 59264 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction