missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ LTB4 Parameter Assay Kit

Product Code. 18762064
Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18762064

Brand: R&D Systems™ KGE006B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LTB4 Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The LTB4 Parameter Assay Kit quantifies multi-species Leukotriene B4/LTB4 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 10.30 to 2500 pg/mL
Assay Sensitivity 10.9 pg/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity LTB4 Levels in Various Samples
Sufficient For 1 x 96-well Plate
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.