missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LTA4H Monoclonal Antibody (24GB5540)
GREENER_CHOICE

Mouse Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen LTA4H
Clone 24GB5540
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274996
GREENER_CHOICE
View Documents
Invitrogen™
MA557442
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276395
View Documents
Invitrogen™
MA557443
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LTA4H Monoclonal Antibody for Western Blot, Flow

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications

Specifications

LTA4H
-20°C
Monoclonal
Liquid
IgG2b
Human, Mouse, Rat
LTA4H
Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27
LTA4H
Primary
Affinity chromatography
24GB5540
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09960, P24527, P30349
16993, 299732, 4048
Recombinant protein of human LTA4H (aa 1-611).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.