Learn More
Invitrogen™ LTA4H Monoclonal Antibody (24GB5535)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557441
 View more versions of this product
Product Code. 30275482
  • £172.00 / 20µL
Description
LTA4H Monoclonal Antibody for Western Blot
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LTA4H
Monoclonal
Unconjugated
LTA4H
Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27
Mouse
Affinity chromatography
RUO
299732, 4048
-20°C
Liquid
Flow Cytometry, Western Blot
24GB5535
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09960, P30349
LTA4H
Recombinant protein of human LTA4H (aa 1-611).
20 μL
Primary
Human, Rat
Antibody
IgG2b