Learn More
LST1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21361-1-AP-150UL
 View more versions of this product
Product Code. 17281366
  • £349.00 / 150µL
Description
The LST1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LST1. This antibody recognizes human, mouse antigen. The LST1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
LST1
Polyclonal
Unconjugated
150 μL
7940
Liquid
Western Blot, ELISA
450 μg/mL
Rabbit
Primary
Human, Mouse