Learn More
Invitrogen™ LST1 Monoclonal Antibody (LST1/02)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110400
Description
This antibody reacts with an extracellular epitope of LST1, an approximately 6-11 kDa protein expressed as various transmembrane or soluble forms. LST1 is found predominantly on monocytes and dendritic cells. It migrates on SDS PAGE gels as approximately 25-28 kDa molecule.
LST1 (leukocyte-specific transcript, also known as B144) is expressed in cells of myeloid/erythroid lineage (monocytes, granulocytes, dendritic cells, platelets, erythrocytes). At least 14 alternatively spliced variants (LST1/A - LST1/N) can be detected; some of them (LST1/A, B, C, G, I, K) are transmembrane cell surface-exposed forms, the other are soluble. LST1 induces production of long, thin filopodia in dendritic cells, has an inhibitory effect on lymphocyte proliferation and may have an immunomodulatory role. LST1/A is an 11 kDa transmembrane adaptor present in membrane rafts and forms spontaneously covalent homodimers. Its intracellular domain contains two tyrosine motifs, one of them being an ITIM very similar to such motifs in Siglec.
Specifications
| LST1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| O00453 | |
| LST1 | |
| KLH-fused peptide corresponding to amino acids SSEGPDLRGRDKRGT of human LST1 (aa 66-80). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| LST1/02 | |
| Unconjugated | |
| LST1 | |
| B144; D6S49E; DAAP-90L16.2; leukocyte specific transcript 1; leukocyte-specific transcript 1 protein; LST1; LST-1; lymphocyte antigen 117; MGC119006; MGC119007; Protein B144 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7940 | |
| 4°C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.