missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LST1 Antibody (LST1/02), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-45072CL1
This item is not returnable.
View return policy
Description
LST1 Monoclonal antibody specifically detects LST1 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| LST1 | |
| Monoclonal | |
| CoraFluor 1 | |
| B144D6S49ELST-1, leucocyte specific transcript 1, leukocyte specific transcript 1, leukocyte-specific transcript 1 protein, lymphocyte antigen 117, MGC119006, MGC119007, Protein B144 | |
| KLH-fused peptide corresponding to amino acids SSEGPDLRGRDKRGT of human LST1 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 7940 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| LST1/02 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction