missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115524
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LSS.
The protein encoded by this gene catalyzes the conversion of (S)-2, 3 oxidosqualene to lanosterol. The encoded protein is a member of the terpene cyclase/mutase family and catalyzes the first step in the biosynthesis of cholesterol, steroid hormones, and vitamin D. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| LSS | |
| Polyclonal | |
| Unconjugated | |
| LSS | |
| 2,3-epoxysqualene-lanosterol cyclase; 2,3-epoxysqualene--lanosterol cyclase; 2,3-oxidosqualene: lanosterol cyclase; 2810025N20Rik; CTRCT44; D10Ertd116e; hOSC; lanosterol synthase; lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase); LSS; OSC; oxidosqualene lanosterol cyclase; oxidosqualene--lanosterol cyclase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4047 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P48449 | |
| LSS | |
| A synthesized peptide derived from human LSS(Accession P48449), corresponding to amino acid residues A354-R404. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction