missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MTEGTCLRRR GGPYKTEPAT DLGRWRLNCE RGRQTWTYLQ DERAGREQTG LEAYALGLDT KNYFKDLPKA HTAFEGALNG MTF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.
The protein encoded by this gene catalyzes the conversion of (S)-2, 3 oxidosqualene to lanosterol. The encoded protein is a member of the terpene cyclase/mutase family and catalyzes the first step in the biosynthesis of cholesterol, steroid hormones, and vitamin D. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Antigen | LSS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LSS |
| Gene Accession No. | P48449 |
| Gene Alias | 2,3-epoxysqualene-lanosterol cyclase; 2,3-epoxysqualene--lanosterol cyclase; 2,3-oxidosqualene: lanosterol cyclase; 2810025N20Rik; CTRCT44; D10Ertd116e; hOSC; lanosterol synthase; lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase); LSS; OSC; oxidosqualene lanosterol cyclase; oxidosqualene--lanosterol cyclase |
| Gene Symbols | LSS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
