missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSS, Mouse anti-Human, Clone: 2E4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004047-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: FESCEERRYLQSAQSQIHNTCWAMMGLMAVRHPDIEAQERGVRCLLEKQLPNGDWPQENIAGVFNKSCAISYTSYRNIFPIWALGRFSQLYPERALAGHPSpecifications
| LSS | |
| Monoclonal | |
| Unconjugated | |
| lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | |
| FLJ25486/FLJ35015/FLJ39450/FLJ46393/OSC | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2E4 | |
| In 1x PBS, pH 7.4 | |
| NM_001001438 | |
| LSS | |
| LSS (NP_001001438.1, 633 a.a. ∼ 732 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4047 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction