missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | LSS |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229109
|
Novus Biologicals
NBP3-38351-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228252
|
Novus Biologicals
NBP3-38351-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LSS Polyclonal antibody specifically detects LSS in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| LSS | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| EC 5.4.99.7, FLJ350152,3-epoxysqualene--lanosterol cyclase, FLJ39450, FLJ46393, hOSC, human lanosterol synthase, EC 5.4.99.710EC 5.4.99, lanosterol synthase, lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)2,3-epoxysqualene-lanosterol cyclase, OSCFLJ25486, Oxidosqualene--lanosterol cyclase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human LSS (NP_002331.3).,, Sequence:, MTEGTCLRRRGGPYKTEPATDLGRWRLNCERGRQTWTYLQDERAGREQTGLEAYALGLDTKNYFKDLPKAHTAFEGALNGMTFYVGLQAEDGHWTGDYGGPLFLLPGLLITCHVARIPLPAGYREEIVRYLRSVQLPDGGWGLHIEDKSTVFGTALNYVSLRILGVGPDDPDLVRARNILHKKGGAVAIPSWGKFWLAVLNVYSWEGLNTLFPEMWLFPDWAPAHPSTLWCHCRQVYLPMSYCYAVRLSAAEDPLVQSLRQELYVEDFASIDWLAQRNNVAPDELYTPHSWLLRVVYALL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100 - 1:500, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 4047 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title